missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WIPI2 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82059F
This item is not returnable.
View return policy
Description
WIPI2 Polyclonal antibody specifically detects WIPI2 in Human, Rat samples. It is validated for ELISA
Specifications
| WIPI2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| ELISA | |
| FITC | |
| ATG18B, Atg21, CGI-50, DKFZP434J154, DKFZp686P02188, FLJ12979, FLJ14217, FLJ42984, WD repeat domain phosphoinositide-interacting protein 2, WD repeat domain, phosphoinositide interacting 2, WD40 repeat protein interacting with phosphoinositides 2, WIPI-2, WIPI49-like protein 2 | |
| Rabbit polyclonal WIPI2 antibody was raised against a 17 amino acid peptide near the amino terminus of human WIPI2. The immunogen is located within the first 50 amino acids of WIPI2. Amino Acid Squence: CANFNQDNTEVKGASRAA | |
| 0.1 mL | |
| Autophagy, Cell Biology, Endocrinology | |
| 26100 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction