Learn More
Wuertt Celluloid™ Wire Beschic Test Tube Rack
Product Code. p-8008801
Click to view available options
Array:
2 x 6
4 x 6
Dimensions (L x W x H):
50 x 130 x 70 mm
90 x 130 x 70 mm
Unit Size:
Each
Product Code. 10282051
Brand: Wuertt Celluloid™ 5272615
Only null left
Add to basket
Edge
Add to basket
Specifications
Product Type Test Tube Rack
Array 2 x 6
Material Wire
Positions 12
For Use With (Application) 4 x 6 Glasses, nylon
Quantity 1 Each
Temperature (Metric) Range 80°C
Dimensions (L x W x H) 50 x 130 x 70 mm
Length (Metric) 50 mm
Width (Metric) Slide 130 mm
Height (Metric) 70 mm