Learn More
TFA Dostmann Wireless Thermometer LOGOneo
Description
Always in the know with the temperature – with the LOGOneo wireless thermometer from TFA. The slim instrument receives the outdoor temperature on site by a separate wireless transmitter; an internal sensor measures the indoor temperature. The maximum and minimum values are stored and displayed at the touch of a button. Optionally, the values can be reset automatically every day. Controlling the room temperature is an important factor for the creation of a comfortable and healthy living environment and it helps save energy. Even one degree of temperature reduction can save up to 6% on heating costs. With the outdoor temperature, you get reliable and important information for your home, garden, leisure and work. In addition, the device has a memory for highest and lowest values and a display for the time. Two more transmitters for temperature can be connected, thus allowing the monitoring of other rooms, e.g. in the nursery, bedroom, cellar or garage.
- Wireless transmission of outdoor temperature
- Indication of indoor temperature
- Maximum and minimum values with manual or auto-reset
- Display of the time
- Expandable up to 3 transmitters
Specifications
Specifications
| Material | Plastic |
| Calibrated | Yes |
| Display | LCD |