Learn More
TFA Dostmann Wireless Thermometer LOGOneo
Product Code. 17901322
Click to view available options
Unit Size:
Each
Product Code. 17901322
Brand: TFA Dostmann 30.3071.54
Only null left
Add to basket
Add to basket
The wireless thermometer LOGOneo shows the time, the indoor and outdoor temperature. The maximum and minimum values are stored and displayed at the touch of a button. Two more transmitters for temperature can be connected, thus allowing the monitoring of other rooms.
Always in the know with the temperature – with the LOGOneo wireless thermometer from TFA. The slim instrument receives the outdoor temperature on site by a separate wireless transmitter; an internal sensor measures the indoor temperature. The maximum and minimum values are stored and displayed at the touch of a button. Optionally, the values can be reset automatically every day. Controlling the room temperature is an important factor for the creation of a comfortable and healthy living environment and it helps save energy. Even one degree of temperature reduction can save up to 6% on heating costs. With the outdoor temperature, you get reliable and important information for your home, garden, leisure and work. In addition, the device has a memory for highest and lowest values and a display for the time. Two more transmitters for temperature can be connected, thus allowing the monitoring of other rooms, e.g. in the nursery, bedroom, cellar or garage.
  • Wireless transmission of outdoor temperature
  • Indication of indoor temperature
  • Maximum and minimum values with manual or auto-reset
  • Display of the time
  • Expandable up to 3 transmitters
Specifications
Material Plastic
Calibrated Yes
Display LCD