Learn More
Witeg™ Witopet™ Variable Microliter Pipetter
Draw and dispense with precision using this economically priced micropipetter. Witeg™ Witopet™ Variable Microliter Pipetter is German-made, highly accurate, color-coded, and ergonomically designed.
Brand: Witeg™ 5 402 002
Additional Details : Weight : 0.80000kg
Description
- Variable volume (range).
- Economical.
- Ergonomic design.
- Lightweight.
- Color-coded
- Robust construction.
- Accepts all established brands of tips.
- Autoclavable.
Specifications
0.08uL | |
All established brands. | |
Red | |
1 | |
Pipette | |
Universal |
DIN EN ISO 8655 | |
Autoclavable | |
Universal | |
Witopet™ | |
0.2 to 2.0 μL |