Learn More
WNK1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B6A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68149-20UL
 View more versions of this product
Product Code. 18087045
  • € 145.00 / 20µL
Description
The WNK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WNK1. This antibody recognizes human, mouse, rat antigen. The WNK1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
WNK1
1B6A10
CoraLite Plus 488
Mouse
Primary
Human, Mouse, Rat
155 kDa
IgG2a
Flow Cytometry (Intracellular Staining)
1000 μg/mL
Erythrocyte 65 kDa protein, HSAN2, HSN2, hWNK1, KDP, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1, PSK, WNK1
20 μL
65125
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.