Learn More
WNK1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1B6A10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68149-20UL
Description
The WNK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human WNK1. This antibody recognizes human, mouse, rat antigen. The WNK1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications
| WNK1 | |
| 1B6A10 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 155 kDa | |
| IgG2a |
| Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| Erythrocyte 65 kDa protein, HSAN2, HSN2, hWNK1, KDP, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1, PSK, WNK1 | |
| 20 μL | |
| 65125 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |