missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human WNK2, GST Tag Recombinant Protein

Product Code. 11543657
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
This item is not returnable. View return policy

Product Code. 11543657

Brand: Invitrogen™ PV4341

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic.

WNK2 is a serine/threonine kinase involved in signal transduction and cellular communication. WNK kinases contain a lysine upstream of the traditional position, within a glycine string. This lysine functions as an anchor and orients ATP through interactions with the alpha and beta phosphoryl groups. The catalytic domains of WNK2, WNK3 and WNK4 are 95% homologous to WNK1. The human WNK1 gene encodes a 2,382 amino acid protein that is primarily expressed in heart, kidney, muscle and distal nephron. The human WNK3 gene encodes a protein that is primarily expressed in brain; the human WNK4 gene encodes a 1,243 amino acid protein that is expressed in kidney. Aberrant function of WNK kinases and their associated signaling pathways are implicated in hypertension, increased renal salt reabsorption and impaired K+ and H+ excretion.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y3S1
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.5mM EDTA, 150mM NaCl, 50% glycerol, 0.02% Triton X-100, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 65268
Molecular Weight (g/mol) 64.9 kDa
Name Human WNK2, GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1810073P09Rik; Antigen NY-CO-43; AW122246; ESTM15; Kiaa1760; kinase deficient protein; LOW QUALITY PROTEIN: serine/threonine-protein kinase WNK2; mitogen-activated protein kinase kinase kinase; mKIAA1760; NY-CO-43; P/OKcl.13; PRKWNK2; protein kinase lysine-deficient 2; Protein kinase with no lysine 2; protein kinase, lysine deficient 2; RGD1307284; SDCCAG43; Serine/threonine-protein kinase WNK2; serologically defined colon cancer antigen 43; WNK lysine deficient protein kinase 2; Wnk2; X83337
Common Name WNK2
Gene Symbol WNK2
KinaseFamily WNK Kinase Family
Protein Length 166-489
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence 166-489
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
Target Entry WNK2
Target Gene WNK lysine deficient protein kinase 2
Expression System Baculovirus
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Catalytic
Form Liquid
Protein Subtype Serine⁄Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.