missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5b Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41306AF350
This item is not returnable.
View return policy
Description
Wnt-5b Polyclonal antibody specifically detects Wnt-5b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Wnt-5b | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 350 | |
| MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein | |
| Antibody was raised against a 15 amino acid peptide near the center of human WNT5B. The immunogen is located within amino acids 130 - 180 of WNT5. Amino Acid Squence: CSRTARPKDLPRDWL | |
| 0.1 mL | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 81029 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction