Learn More
Wnt-5b Antibody (SR1784), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-22311-100ul
 View more versions of this product
Product Code. 18620755
  • € 449.00 / 100µL
Description
Wnt-5b Monoclonal antibody specifically detects Wnt-5b in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wnt-5b
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, Flow Cytometry
SR1784
Western Blot 1:500-1:2000, Flow Cytometry 1:50
MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein
A synthesized peptide derived from human Wnt-5b (Uniprot #: Q9H1J7)
100 μg
Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
81029
Store at -20° C. Avoid freeze/thaw cycles.