missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNT10A, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova PAB13043.100ug
This item is not returnable.
View return policy
Specifications
| WNT10A | |
| Polyclonal | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| wingless-type MMTV integration site family, member 10A | |
| WNT10A | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human WNT10A. | |
| RUO | |
| 80326 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| In PBS (0.02% sodium azide) | |
| FLJ14301 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction