missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT10B Polyclonal Antibody
GREENER_CHOICE

Product Code. 15951125
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15951125 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15951125 Supplier Invitrogen™ Supplier No. PA572461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WNT10B Polyclonal Antibody for Western Blot, IHC (P), Flow

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It may be involved in breast cancer, and its protein signaling is likely a molecular switch that governs adipogenesis. This protein is 96% identical to the mouse Wnt10b protein at the amino acid level. This gene is clustered with another family member, WNT1, in the chromosome 12q13 region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT10B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene WNT10B
Gene Accession No. O00744, P48614
Gene Alias Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12
Gene Symbols WNT10B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 193-222 amino acids from the Central region of human WNT10B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22410, 7480
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.