missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT11 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15959294
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15959294 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15959294 Supplier Invitrogen™ Supplier No. PA566600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ATKVVHRPMGT RKHLVPKDLD IRPVKDSELV YLQSSPDFCM KNEKVGSHGT QDRQCNKTSN GSDSCD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WNT11
Gene Accession No. O96014
Gene Alias HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11
Gene Symbols WNT11
Host Species Rabbit
Immunogen Recombinant Human WNT11. Recombinant protein control fragment (Product #RP-107254).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7481
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.