missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt16 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 213.00 - € 507.00
Specifications
| Antigen | Wnt16 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30227209
|
Novus Biologicals
NBP3-33220-100ul |
100 μL |
€ 507.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228522
|
Novus Biologicals
NBP3-33220-20ul |
20 μL |
€ 213.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wnt16 Monoclonal antibody specifically detects Wnt16 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| Wnt16 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Wnt Signaling Pathway | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 51384 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| protein Wnt-16, wingless-type MMTV integration site family, member 16 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Wnt16 (Q9UBV4).,, Sequence:, MDRAALLGLARLCALWAALLVLFPYGAQGNWMWLGIASFGVPEKLGCANLPLNSRQKELCKRKPYLLPSIREGARLGIQECGSQFRHERWNCMITAAATT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title