missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT2 Recombinant Rabbit Monoclonal Antibody (6W3P9)

Product Code. 31903711
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31903711 20 μL 20µL
30131598 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31903711 Supplier Invitrogen™ Supplier No. MA569211

Please to purchase this item. Need a web account? Register with us today!

Rabbit Recombinant Monoclonal Antibody

Immunogen Sequence: GDYLWRKYNG AIQVVMNQDG TGFTVANERF KKPTKNDLVY FENSPDYCIR DREAGSLGTA GRVCNLTSRG MDSCEVMCCG RGYDTSHVTR MTKCGCKFHW.

WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 6W3P9
Concentration 0.66 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Wnt2
Gene Accession No. P09544, P21552
Gene Alias 2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2
Gene Symbols Wnt2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 231-330 of human WNT2
Purification Method Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114487, 22413, 7472
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.