missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT2 Recombinant Rabbit Monoclonal Antibody (HL1999)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550358
This item is not returnable.
View return policy
Description
WNT2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.
Specifications
| WNT2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P09544, P21552 | |
| Wnt2 | |
| Recombinant fragment of human Wnt2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1999 | |
| Unconjugated | |
| Wnt2 | |
| 2610510E18Rik; etID309852.18; INT1L 1; Int1l1; int-1-like protein 1; INT-1-related protein; IRP; IRP protein; Mirp; protein Wnt-2; secreted growth factor; wingless-related MMTV integration site 2; wingless-type MMTV integration site 2; wingless-type MMTV integration site family member 2; wingless-type MMTV integration site family, member 2; Wnt; WNT 2; Wnt family member 2; Wnt2; Wnt-2; Wnt2a; ZfWnt2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 114487, 22413, 7472 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction