missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt3 Antibody (OTI1G7), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74869DL594
This item is not returnable.
View return policy
Description
Wnt3 Monoclonal antibody specifically detects Wnt3 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Wnt3 | |
| Monoclonal | |
| DyLight 594 | |
| INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein | |
| Recombinant protein fragment corresponding to amino acids 128-355 of human Wnt3 (NP_110380) produced in E.coli. | |
| 0.1 mL | |
| Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 7473 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| OTI1G7 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction