missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578226
This item is not returnable.
View return policy
Description
Positive Control: Wnt3-transfected 293T cells, Want3A-transfected 293T cells Predicted Reactivity: Mouse (99%), Rat (99%), Zebrafish (91%), Chicken (98%), Bovine (99%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WNT3, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
| WNT3 | |
| Polyclonal | |
| Unconjugated | |
| WNT3 | |
| INT4; Int-4; MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4; proto-oncogene Int-4 homolog; proto-oncogene protein Wnt-3; proto-oncogene Wnt-3; TETAMS; wingless-related MMTV integration site 3; Wingless-type MMTV integration site 3, homolog; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; Wnt family member 3; Wnt3; Wnt-3; WNT-3 proto-oncogene protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7473 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.3 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P56703 | |
| WNT3 | |
| Recombinant protein encompassing a sequence within the center region of human WNT3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction