missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT5A Recombinant Rabbit Monoclonal Antibody (JU30-30)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA541244
This item is not returnable.
View return policy
Description
WNT5A Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.
Specifications
| WNT5A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P41221 | |
| WNT5A | |
| Synthetic peptide within human wnt5a aa 331-380. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JU30-30 | |
| Unconjugated | |
| WNT5A | |
| 8030457G12Rik; hWNT5A; OTTHUMP00000212837; OTTHUMP00000212861; Protein Wnt-5a; wingless-related MMTV integration site 5A; wingless-type MMTV integration site 5A; wingless-type MMTV integration site family member 5A; wingless-type MMTV integration site family, member 5A; Wnt family member 5A; WNT5A; Wnt-5a; WNT-5A protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction