missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT7A Polyclonal Antibody

Product Code. 15305191
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15305191 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15305191 Supplier Invitrogen™ Supplier No. PA534329

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

WNT7A Polyclonal Antibody for IHC (P)

Wnt-7a belongs to the Wnt family of signaling proteins that play a key role in maintaining the integrity of embryonic and adult tissues. It is expressed in placenta, kidney, testis, uterus, fetal lung, and fetal and adult brain. Most Wnt proteins can signal though a mechanism called the canonical Wnt pathway, in which Wnt proteins bind to and activate seven-pass transmembrane receptors of the Frizzled family, ultimately leading to the disruption of β-catenin degradation. Intracellular accumulation of β-catenin increases translocation of the protein into the nucleus, where it binds to TCF/LEF transcription factors to induce the expression of numerous genes. Increased Wnt/β-catenin signaling is associated with tumorigenesis in a diverse set of human cancers. However, Wnt-7a/Frizzled-9 signaling has been shown to act as a tumor suppressor in non-small cell lung cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT7A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Wnt7a
Gene Accession No. O00755, P24383
Gene Alias AI849442; postaxial hemimelia; protein Wnt-7a; proto-oncogene Wnt7a protein; px; tw; wingless-related MMTV integration site 7A; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family, member 7A; Wnt family member 7A; WNT7A; Wnt-7a
Gene Symbols Wnt7a
Host Species Rabbit
Immunogen Synthetic 12 amino acid peptide from internal region of human WNT7A.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100055450, 100153445, 100355697, 100719106, 100772978, 114850, 22421, 374168, 533782, 607180, 7476
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Chicken, Guinea Pig, Frog, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.