missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WNT7A Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA596320

Product Code. 16539161

  • € 526.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Samples: Mouse kidney; Cellular Location: extracellular matrix, extracellular space Immunogen sequence: KEKQGQYHRD EGWKWG.

Wnt-7a belongs to the Wnt family of signaling proteins that play a key role in maintaining the integrity of embryonic and adult tissues. It is expressed in placenta, kidney, testis, uterus, fetal lung, and fetal and adult brain. Most Wnt proteins can signal though a mechanism called the canonical Wnt pathway, in which Wnt proteins bind to and activate seven-pass transmembrane receptors of the Frizzled family, ultimately leading to the disruption of β-catenin degradation. Intracellular accumulation of β-catenin increases translocation of the protein into the nucleus, where it binds to TCF/LEF transcription factors to induce the expression of numerous genes. Increased Wnt/β-catenin signaling is associated with tumorigenesis in a diverse set of human cancers. However, Wnt-7a/Frizzled-9 signaling has been shown to act as a tumor suppressor in non-small cell lung cancers.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WNT7A
Polyclonal
Unconjugated
Wnt7a
AI849442; postaxial hemimelia; protein Wnt-7a; proto-oncogene Wnt7a protein; px; tw; wingless-related MMTV integration site 7A; wingless-type MMTV integration site 7A; wingless-type MMTV integration site family member 7A; wingless-type MMTV integration site family, member 7A; Wnt family member 7A; WNT7A; Wnt-7a
Rabbit
Affinity Chromatography
RUO
22421, 7476
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.28 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
O00755, P24383
Wnt7a
A synthetic peptide corresponding to a sequence within amino acids 101-200 of human WNT7A (NP_0046162).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.