missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT9A Recombinant Rabbit Monoclonal Antibody (HL2422)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550524
This item is not returnable.
View return policy
Description
WNT9A Recombinant Monoclonal Antibody for Western Blot, IHC (P)
WNT9A is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. WNT9A is likely to signal over only few cell diameters.
Specifications
| WNT9A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O14904, Q8R5M2 | |
| WNT9A | |
| Synthetic peptide of human Wnt9a. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2422 | |
| Unconjugated | |
| WNT9A | |
| MGC138165; MGC141991; Protein Wnt-14; Protein Wnt-9a; wingless related MMTV integration site 9A; wingless-type MMTV integration site 14; wingless-type MMTV integration site 9A; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A; Wnt family member 9A; WNT14; wnt-14; Wnt9a | |
| Rabbit | |
| Protein A | |
| RUO | |
| 216795, 287357, 7483 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction