Learn More
Techsit Worktech Laboratory ESD High Chair
Description
- ESD Cleanroom vinyl high chair with adjustable footring on ESD glides
- Black ESD/Anti-static Vinyl that is easy to clean
- Fully adjustable Seat height, back height, and back rake
- Ergonomically designed and contoured seat and back
- Cleanroom class 7 suitable
- BS EN61340-5-1 Conductivity 104 - 106 Ohms
High ESD Vinly Cleanroom Chair
Specifications
Specifications
| Product Type | Cleanroom ESD Chair |
| Color | Black |
| Autoclavable | Not Autoclavable |
| Back Style | Ergonomic |
| Style | High back |
| With Armrest | No |
| With Backrest | Yes |
| With Casters | No |
| Footrest | Yes |
| With Lumbar Support | Yes |
| Show More |