Learn More
B Braun™ Wound and Bladder Syringe
Click to view available options
Volume (Metric):
100 mL
Unit Size:
Pack of 100
Description
- Smooth plunger action and double-seal stopper
- High visibility – transparent barrel with bold graduations
- Secure backstop to prevent accidental plunger withdrawal
- Double seal stopper to prevent medication leakage
- Large, stable finger grips
- DEHP-free and latex-free
Specifications
Specifications
| Material | Polyisoprene Rubber (Tip), Polypropylene (Barrel) |
| Product Type | Syringe |
| Packaging | 50/Cs. |
| Volume (Metric) | 100 mL |