missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRCH1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13230
This item is not returnable.
View return policy
Description
WRCH1 Polyclonal specifically detects WRCH1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| WRCH1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| ARHU, CDC42L1GTP-binding protein-like 1, DJ646B12.2, fJ646B12.2, FLJ10616,2310026M05Rik, G28K, GTP-binding protein like 1, GTP-binding protein SB128, hG28K, ras homolog gene family, member U, ras-like gene family member U, Rho GTPase-like protein ARHU, rho-related GTP-binding protein RhoU, Ryu GTPase, Wnt-1 responsive Cdc42 homolog 1, WRCH-1, WRCH1CDC42-like GTPase 1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RHOU | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LREDVKVLIELDKCKEKPVPEEAA | |
| 0.1 mL | |
| Cell Biology, Signal Transduction | |
| 58480 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction