Learn More
Cryo-Industrial™ Wrist Fabric Gloves
Catalog Number 11883851
Click to view available options
Size:
Large
X-Large
Cryo-Industrial™ Wrist Fabric Gloves provide protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
  • Protection for hands and arms when working in hazardous, ultra-cold environments down to -200°C
  • Gloves are designed to provide a high level of thermal protection and abrasion resistance for heavy-duty applications
  • Wrist length
  • Made from state-of-the-art materials
  • Designed to keep the user dry and to wick moisture away from your hands
  • Can be worn in comfort for extended periods of time.
  • Multi-layered construction allows for a maximum level of thermal protection, flexibility, and dexterity
Color Blue
Disposable Reusable
Material Fabric
Powdered Powder Free
Product Type Cryogenic Gloves
Size X-Large
Sterility Non-sterile
Style Wrist-length