missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WRN Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-472R
This item is not returnable.
View return policy
Description
WRN Polyclonal antibody specifically detects WRN in Human, Mouse samples. It is validated for Western Blot
Specifications
| Western Blot | |
| 50mM Sodium Borate | |
| Immunogen affinity purified | |
| Human | |
| Purified |
| DyLight 550 | |
| DKFZp686C2056, DNA helicase, RecQ-like type 3, EC 3.1, EC 3.6.1, EC 3.6.4.12, Exonuclease WRN, RecQ protein-like 2, RecQ3, RECQ3RECQL3, RECQL2Werner syndrome ATP-dependent helicase, Werner syndrome, Werner syndrome, RecQ helicase-like | |
| 0.1 mL | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction