missing translation for 'onlineSavingsMsg'
Learn More

WS6, MedChemExpress

Product Code. 16481397
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16481397

Brand: Medchem Express HY12461

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WS6 is a novel small molecule that promotes β cell proliferation in rodent and human primary islets with EC50 of 0.28 uM(R7T1 cell viability).

WS6 is a novel small molecule that promotes β cell proliferation in rodent and human primary islets with EC50 of 0.28 uM(R7T1 cell viability).

Specifications

Chemical Name or Material WS6
CAS 1421227-53-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31F3N6O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (175.87 mM)
SMILES O=C(NC1=CC=C(CN2CCN(C)CC2)C(C(F)(F)F)=C1)CC3=CC=C(OC4=NC=NC(NC(C5CC5)=O)=C4)C=C3
Molecular Weight (g/mol) 568.59
Formula Weight 568.59
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.