missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WSB1 Antibody (K5P2H10*E10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50365IR
This item is not returnable.
View return policy
Description
WSB1 Monoclonal specifically detects WSB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| WSB1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WSB1 | |
| Ovalbumin-conjugated synthetic peptide RRVMPTQEVQE | |
| 0.1 mL | |
| Signal Transduction | |
| 26118 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| K5P2H10*E10 | |
| Western Blot, ELISA, Immunohistochemistry | |
| DKFZp564B0482, SOCS box-containing WD protein SWiP-1, SWIP1DKFZp564A122, WD repeat and SOCS box containing 1, WD repeat and SOCS box-containing 1, WD repeat and SOCS box-containing protein 1, WSB-1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction