missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (960522), Alexa Fluor 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB5729AF350
This item is not returnable.
View return policy
Description
WT1 Monoclonal specifically detects WT1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
| WT1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| WT1 | |
| E. coli-derived recombinant human WT1, Met127-Gly249, Accession No. P19544 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 960522 | |
| Flow Cytometry, Immunocytochemistry | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction