missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (WT1/1434R), Alexa Fluor™ 350 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-11622AF350
This item is not returnable.
View return policy
Description
WT1 Monoclonal antibody specifically detects WT1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| WT1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| Recombinant human full-length WT1 protein (Uniprot: P19544 ) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| WT1/1434R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction