missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WT1 Antibody (WT1/857 + 6F-H2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47858IR
This item is not returnable.
View return policy
Description
WT1 Monoclonal specifically detects WT1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| WT1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| WT1 | |
| Recombinant full length human WT1 protein (WT1/857); Recombinant fragment aa1-181 of human WT1 (6F-H2) (Uniprot: P19544) | |
| 0.1 mL | |
| Primary | |
| Recognizes a 47-55kDa-tumor suppressor protein, identified as Wilms tumor and mesothelioma. WT1 protein has been identified in proliferative mesothelial cells, malignant mesothelioma, ovarian carcinoma, gonadoblastoma, nephroblastoma, and desmoplastic small round cell tumor. Lung adenocarcinomas rarely stain positive with this antibody. WT1 protein expression in mesothelial cells has become a reliable marker for the diagnosis of mesotheliomas. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| WT1/857 + 6F-H2 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence | |
| AWT1, NPHS4GUD, WAGR, Wilms tumor 1, Wilms tumor protein, WIT-2, WT33 | |
| Mouse | |
| Protein A or G purified | |
| Angiogenesis, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Tumor Suppressors | |
| 7490 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction