missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Antibody (PCRP-WTAP-1A4), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23995PCP
This item is not returnable.
View return policy
Description
WTAP Monoclonal antibody specifically detects WTAP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin)
Specifications
| WTAP | |
| Monoclonal | |
| PerCP | |
| Female-lethal(2)D homolog, hFL(2)D, KIAA0105DKFZp686F20131, MGC3925, PNAS-132, pre-mRNA-splicing regulator WTAP, putative pre-mRNA splicing regulator female-lethal(2D), Wilms tumor 1 associated protein, Wilms tumor 1-associating protein, Wilms' tumour 1-associating protein, WT1-associated protein | |
| Recombinant full-length human WTAP protein | |
| 0.1 mL | |
| Cancer, Tumor Suppressors | |
| 9589 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin) | |
| PCRP-WTAP-1A4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction