missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WTAP Monoclonal Antibody (OTI12G6)
GREENER_CHOICE

Product Code. 15986564
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15986564 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15986564 Supplier Invitrogen™ Supplier No. MA526977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WTAP Monoclonal Antibody for Western Blot, IHC (P)

Wilms' tumor (WT) is an embryonal malignancy of the kidney that affects 1 in 10,000 infants and is observed in both sporadic and inherited forms. The Wilms' tumor protein (WT1) binds the DNA sequence GCGGGGGCG, a recognition element common to the early growth response (Egr) family of Zn2+ finger transcriptional activators, and functions as a transcriptional repressor. WTAP (Wilms tumor 1-associating protein) is a ubiquitously expressed nuclear protein that interacts with WT1 and may be involved in regulating mRNA splicing. WTAP is found in nuclear speckles, where it regulates the G2/M cell cycle transition by binding to the 3' UTR of cyclin A2, thus enhancing its stability. Additionally, WTAP inhibits expression of WT1 target genes and is able to impair the ability of WT1 to bind DNA. Two isoforms of WTAP exist due to alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WTAP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI12G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WTAP
Gene Accession No. Q15007
Gene Alias 2810408K05Rik; 9430038B09Rik; Female-lethal(2)D homolog; hFL(2)D; KIAA0105; Mum2; PNAS-132; pre-mRNA-splicing regulator WTAP; putative pre-mRNA splicing regulator female-lethal(2D); RGD1563824; Wilms tumor 1 associated protein; wilms tumor 1-associating protein; Wilms' tumour 1-associating protein; Wilms tumour 1-associating protein; WT1-associated protein; WTAP
Gene Symbols WTAP
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-106 of WTAP produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9589
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.