missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTAP Monoclonal Antibody (OTI6C3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808497
This item is not returnable.
View return policy
Description
The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.Specifications
| WTAP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15007 | |
| WTAP | |
| Human recombit protein fragment corresponding to amino acids 2-106 of human WTAP produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6C3 | |
| Unconjugated | |
| WTAP | |
| Mum2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9589 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction