missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWP1 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-49712V
This item is not returnable.
View return policy
Description
WWP1 Polyclonal antibody specifically detects WWP1 in Human samples. It is validated for Western Blot
Specifications
| WWP1 | |
| Polyclonal | |
| Western Blot | |
| AIP5Nedd-4-like ubiquitin-protein ligase, atrophin-1 interacting protein 5, Atrophin-1-interacting protein 5, DKFZP434D2111, EC 6.3.2, EC 6.3.2.-, hSDRP1, NEDD4-like E3 ubiquitin-protein ligase WWP1, TGIF-interacting ubiquitin ligase 1, TIUL1, WW domain containing E3 ubiquitin protein ligase 1, WW domain-containing protein 1 | |
| The immunogen for this product maps to a region between residue 200 and 250 of human WW Domain Containing E3 Ubiquitin Protein Ligase 1 using the numbering given in entry NP_008944.1 (GeneID 11059). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 11059 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction