missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWTR1 Monoclonal Antibody (OTI1A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503438
This item is not returnable.
View return policy
Description
WW domain-containing transcription regulator protein 1 (WWTR1) is a transcriptional coactivator which acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor supression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1. In turn, MOB1 phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. WWTR1 enhances PAX8 and NKX2-1/TTF1-dependent gene activation. WWTR1 regulates the nuclear accumulation of SMADS and has a key role in coupling them to the transcriptional machinery such as the mediator complex. WWTR1 also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition.Specifications
| WWTR1 | |
| Monoclonal | |
| 0.61 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9EPK5, Q9GZV5 | |
| WWTR1 | |
| Full length human recombit protein of human WWTR1 produced in HEK293 cell. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI1A12 | |
| Unconjugated | |
| WWTR1 | |
| TAZ | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 25937, 609743, 97064 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction