missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WWTR1 Monoclonal Antibody (OTI1A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503438S
This item is not returnable.
View return policy
Description
WWTR1 Monoclonal antibody specifically detects WWTR1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| WWTR1 | |
| Monoclonal | |
| 0.61 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P05093 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1586 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1A12 | |
| Unconjugated | |
| CYP17A1 | |
| 2310058J06Rik; 2610021I22Rik; C78399; DKFZp586I1419; FLJ27004; FLJ45718; Tafazzin; TAZ; transcriptional coactivator with PDZ binding motif; transcriptional co-activator with PDZ-binding motif; transcriptional coactivator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); WW domain containing transcription regulator 1; WW domain-containing transcription regulator protein 1; Wwtr1 | |
| Full length human recombinant protein of human CYP17A1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction