missing translation for 'onlineSavingsMsg'
Learn More

WZ4141R, MedChemExpress

Product Code. p-200096590
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Product Code. Quantity unitSize
16450989 1 mg 1mg
16446497 5 mg 5mg
16410157 50 mg 50mg
16403778 10 mg 10mg
4 options
This item is not returnable. View return policy

Product Code. 16450989

Brand: Medchem Express HY103016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WZ4141R is an intermediate in the synthesis of compounds.

WZ4141R is an intermediate in the synthesis of compounds.

Specifications

Chemical Name or Material WZ4141R
CAS 2319589-98-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H15N3O2
Quantity 1 mg
SMILES O=C(CC)NC1=CC(OC2=C3C(NC=C3)=NC=C2)=CC=C1
Molecular Weight (g/mol) 281.31
Formula Weight 281.31
Percent Purity 98.03%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.