Learn More
Melford X-Glucoside
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
Description
Histochemical substrate for β-D-Glucosidase.
Histochemical substrate for β-D-Glucosidase.
Specifications
Specifications
| Quantity | 100 mg |