Learn More
Abeba™ X-Light Low Safety Shoe
Product Code. 17553155
Change view
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17553155 35 1 pair
17563155 36 1 pair
17573155 37 1 pair
17583155 38 1 pair
17593155 39 1 pair
17503165 40 1 pair
17513165 41 1 pair
17523165 42 1 pair
17533165 43 1 pair
17543165 44 1 pair
17553165 45 1 pair
17563165 46 1 pair
17573165 47 1 pair
17583165 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 17553155 Supplier Abeba™ Supplier No. 711038/35
Only null left
Add to basket
Add to basket
Abeba X-Light Low Safety Shoe has a breathable inner lining with silver ions.
  • Size 35 to 48
  • CE, EN ISO 20345:2011, S2, SRC
  • Smooth leather, black
  • Breathable lining with Silver ions
  • Instep area with lacing
  • Slip resistance PU outsole sole
  • Steel toe cap
  • Replaceable Active Comfort Insole (art. 351620)
  • Antistatic
  • Reflective material
Specifications
Closure Type Laces
Color Black
Material Leather
Size 35
Toe Material Steel
Certifications/Compliance (Europe) CE, EN ISO 20345:2011, S2, SRC