Learn More
BRAND™ X-Shape PTFE Magnetic Stirring Bars
Product Code. 10598401 Shop All Brand Products
Click to view available options
Diameter (Metric):
10 mm
20 mm
25 mm
30 mm
38 mm
Height (Metric):
5mm
9 mm
10mm
12mm
15mm
Unit Size:
Pack of 10
Product Code. 10598401
Brand: BRAND™ 137632
Only null left
Add to basket
Add to basket
An excellent option for liquid agitation in the laboratory.
  • Steady spinning position
  • Excellent stirring action
  • Provided in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Height (Metric) 9 mm
Coating PTFE
Diameter (Metric) 20 mm
Material PTFE
Quantity 10/Pk.
Shape Cross
Type Magnetic Stir Bar