Learn More
Hellma X1 Fluoroescence cuvette, QS, 30ul, 8.5mm CentreHeight, 1.5x1.5mm Light Path, 3 windows
Brand: Hellma 105-252-85-40
X1 Fluoroescence cuvette, QS, 30ul, 8.5mm CentreHeight, 1.5x1.5mm Light Path, 3 windows