Learn More
Ratiolab X10 Cardboard 1/2 format Grid Insert 8x4 for Cryobox, 124x58x30mm
Brand:  Ratiolab 5120218
X10 Cardboard 1/2 format Grid Insert 8x4 for Cryobox, 124x58x30mm
Product Code. 17453713
  • € 18.35 / Pack of 10
Only null left
In Stock