missing translation for 'onlineSavingsMsg'
Learn More

Chemtronics Coventry Poly Double-Onyx ESD Wipes

Product Code. 17521165 Shop All Texwipe Products
Click to view available options
Dimensions (L x W):
9 x 9 in.
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17521165

Brand: Chemtronics 6259BESD/X10

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

ESD Wipes are made from 2nd generation black no-run monofilament polyester knit, providing excellent strength, absorbency and chemical compatibility.

Features and Benefits

  • Class 100 to 1000 (ISO 5-6) black, laser-sealed edge polyester wipe
  • Static dissipative within 107-109 range for S20.20 compliance
  • Conductive material is double-sided and double-density for maximum continuity to ground
  • High durability for cleaning rough, abrasive, or irregular surfaces
  • Monofilament 'no-run' construction does not release loose fibers
  • Excellent solvent and acid resistance
  • Low ionic, nonvolatile residue and particle contamination
  • Good absorbency
  • Applications
  • Semiconductor Wafer Fabs
  • Aerospace Production Areas
  • Disk Drives Production Areas
  • Pharmaceutical / Biotechnical Production Areas
  • General Clean room Cleaning
  • Dust and Make-up Detection / Inspection
  • Clean static sensitive surfaces

Specifications

Product Type Anti-static Decontamination Wipe
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester
Dimensions (L x W) 9 x 9 in.
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.