Learn More
Edulab Mohr's Clips (Tubing Clamps)
Click to view available options
Type:
Clamp
Unit Size:
Pack of 10
Description
- Pack of 10 Mohr’s Clips suitable for clamping and controlling tubing flow
- Provides a secure hold to prevent leaks during laboratory experiments
- Reusable and easy to attach or remove for flexible use
- Lightweight design for simple handling and reliable performance
Specifications
Specifications
| Type | Clamp |
| Material | Nickel-plated Brass |
| Color | Silver |
| Jaw Type | 2-Prong |