Learn More
Edulab Mohr's Clips (Tubing Clamps)
Product Code. 30280269
Click to view available options
Type:
Clamp
Unit Size:
Pack of 10
Product Code. 30280269
Brand: Edulab CH0290B
Only null left
Add to basket
Add to basket
Durable clips designed for securing tubing in laboratory applications.
  • Pack of 10 Mohr’s Clips suitable for clamping and controlling tubing flow
  • Provides a secure hold to prevent leaks during laboratory experiments
  • Reusable and easy to attach or remove for flexible use
  • Lightweight design for simple handling and reliable performance
Specifications
Type Clamp
Material Nickel-plated Brass
Color Silver
Jaw Type 2-Prong