missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Orange Serum Agar
Click to view available options
Fill Weight:
250 to 254 g
Unit Size:
Pack of 10
Description
Isolate and enumerate spoilage organisms of citrus products with Thermo ScientificTM OxoidTM Orange Serum Agar in ready to use format
Specifications
Specifications
| Color | Straw |
| Content And Storage | 2°C to 25°C |
| Format | 300 mL Sirop Bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar |
| Fill Weight | 250 to 254 g |
| Packaging Type | 10 Sirop Bottles with screw cap closures in a box |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction