missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Anaerobe Blood Agar (WC) with Nalidixic Acid and Vancomycin
Click to view available options
Quantity:
10 Plates
Unit Size:
Pack of 10
Description
Engineered to support the growth of anaerobic organisms while inhibiting unwanted bacteria, this specialized agar medium is your go-to solution for accurate and reliable results.
Specifications
Specifications
| Color | Red |
| Content And Storage | 2°C to 12°C |
| Format | 90 mm |
| Manufacturing Origin | United Kingdom |
| Product Type | Agar Plate |
| Fill Volume | 17 mL |
| Packaging Type | 1 x 10 Plates |
| Quantity | 10 Plates |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction