Learn More
Macherey Nagel X100 Filtration cartridges PTFE (polytetrafluoroethylene) 6 mL, 0.20
Brand:  Macherey Nagel 730570.620
X100 Filtration cartridges PTFE (polytetrafluoroethylene) 6 mL, 0.20
Product Code. 15612976
  • ONLINE EXCLUSIVE PRICE
    € 173.00
    € 172.00 / Pack of 100
    Save € 1.00
    0.58% Off
Only null left
Estimated Shipment: 03-02-2026