Learn More
DiversifiedBiotech TOUGH-SPOTS™ Roll Labels - 1.5 mL Tubes (13 mm)
Click to view available options
Color:
Coffee
Gold
Lavender
Maroon
Orange
Pink
Silver
Unit Size:
Pack of 1000
Description
- Circular cryogenic labels formatted for handwriting
- Tough Spots™ on a roll are easy to write PVC cryogenic labels that accept writing from most lab markers
- Tough Spots™ come in a variety of colors and sizes to ensure the right label for the right job
- Adheres to most plastics (100% virgin PP tubes recommended), glass and metals without cracking, peeling or degrading
- Flexible polyolefin material and acrylic adhesive expand and contract with temperature fluctuations
- Label Dimensions: 0.50 in. (13 mm)
- Roll of 1000
Specifications
Specifications
| Adhesive Type | Peel and Stick, Acrylic |
| Color | Orange |
| Finish | Semi-gloss |
| Format | Roll |
| Material | Polyolefin |
| Print Technology | Laser |
| Product Type | Ultra Reflective Letter Label |
| Sterility | Non-sterile |
| Length (English) | 0.50 in. |
| Width (English) | 0.50 in. |
| Show More |