Learn More
Kinesis X1000 Septa 45' shore A PTFE red/silicone white/PTFE red for screw cap 10mm (pack of 1K)
Brand: Kinesis SEP10-03
X1000 Septa 45' shore A PTFE red/silicone white/PTFE red for screw cap 10mm (pack of 1K)