Learn More
Herenz Heinz X150 LMS-Swab Thio Broth, flocked swab,standard tip, blackscrew cap, 2ml ofThio Broth medium
Brand:  Herenz Heinz 1020312
X150 LMS-Swab Thio Broth, flocked swab,standard tip, blackscrew cap, 2ml ofThio Broth medium
Product Code. 30242809
  • ONLINE EXCLUSIVE PRICE
    € 131.15
    € 79.67 / Pack of 150
    Save € 51.48
    39% Off